Assay Details
Gene Symbol: Eral1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013229.1 | NP_001013247.1 | 6 - 7 | 720 | 73 | |
XM_008768087.1 | XP_008766309.1 | 6 - 7 | 716 | 73 | ||
GenBank mRNA | BC089885.1 | - | 6 - 7 | 720 | 7370040 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
