Assay Details
Gene Symbol: Ube2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022690.2 | NP_073181.1 | 3 - 4 | 546 | 134 | |
GenBank mRNA | AF099093.1 | - | 3 - 4 | 513 | 134 | |
BC086980.1 | - | 3 - 4 | 546 | 7192141 | 134 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
