Assay Details
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114606.1 | NP_001108078.1 | 3 - 4 | 484 | 73 | |
XM_006246509.2 | XP_006246571.1 | 3 - 4 | 2081 | 73 | ||
GenBank mRNA | BC081953.1 | - | 3 - 4 | 457 | 7122794 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
