Assay Details
Gene Symbol: Nlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191924.1 | NP_001178853.1 | 3 - 4 | 1838 | 93 | |
XM_006246945.2 | XP_006247007.1 | 3 - 4 | 1640 | 93 | ||
GenBank mRNA | FQ225006.1 | - | 3 - 4 | 693 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
