Assay Details
Gene Symbol: Nlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191924.1 | NP_001178853.1 | 9 - 10 | 2620 | 81 | |
XM_006246945.2 | XP_006247007.1 | 9 - 10 | 2422 | 81 | ||
GenBank mRNA | FQ225006.1 | - | 8 - 9 | 1265 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
