Assay Details
Gene Symbol: Aldh3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031731.2 | NP_113919.2 | 1 - 2 | 269 | 186 | |
XM_006246529.2 | XP_006246591.1 | 1 - 2 | 767 | 186 | ||
GenBank mRNA | FQ210485.1 | - | 1 - 2 | 332 | 186 | |
FQ221576.1 | - | 1 - 2 | 291 | 186 | ||
M73714.1 | - | 1 - 2 | 275 | 186 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
