Assay Details
Gene Symbol: Aldh3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031731.2 | NP_113919.2 | 5 - 6 | 917 | 117 | |
XM_006246529.2 | XP_006246591.1 | 5 - 6 | 1415 | 117 | ||
GenBank mRNA | CB727458.1 | - | 3 - 4 | 380 | 117 | |
FQ210485.1 | - | 5 - 6 | 980 | 117 | ||
FQ221576.1 | - | 5 - 6 | 939 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
