Assay Details
Gene Symbol: Tbc1d16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247938.2 | XP_006248000.1 | 1 - 2 | 389 | 171 | |
XM_008768486.1 | XP_008766708.1 | 2 - 3 | 2687 | 171 | ||
XM_221188.8 | XP_221188.5 | 1 - 2 | 389 | 171 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
