Assay Details
Gene Symbol: Cxcl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017478.1 | NP_001017478.1 | 4 - 5 | 980 | 84 | |
GenBank mRNA | BC079312.1 | - | 4 - 5 | 980 | 84 | |
DQ025528.1 | - | 4 - 5 | 1256 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
