Assay Details
Gene Symbol: Tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107012.1 | NP_001100482.1 | 1 - 2 | 214 | 108 | |
XM_006246704.2 | XP_006246766.1 | 2 - 3 | 1672 | 108 | ||
XM_006246705.2 | XP_006246767.1 | 3 - 4 | 747 | 108 | ||
XM_006246707.2 | XP_006246769.1 | 3 - 4 | 417 | 108 | ||
XM_006246709.2 | XP_006246771.1 | 2 - 3 | 594 | 108 | ||
XM_008767827.1 | XP_008766049.1 | 2 - 3 | 1123 | 108 | ||
XM_008767828.1 | XP_008766050.1 | 3 - 4 | 511 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
