Assay Details
Gene Symbol: Mpdu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107011.1 | NP_001100481.1 | 4 - 5 | 422 | 135 | |
GenBank mRNA | BC087071.1 | - | 4 - 5 | 599 | 135 | |
FQ213141.1 | - | 4 - 5 | 398 | 135 | ||
FQ220130.1 | - | 4 - 5 | 386 | 135 | ||
FQ229625.1 | - | 2 - 3 | 191 | 135 | ||
FQ229903.1 | - | 2 - 3 | 191 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
