Assay Details
Gene Symbol: Slc9a3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053811.3 | NP_446263.2 | 6 - 7 | 927 | 121 | |
XM_006245893.2 | XP_006245955.1 | 5 - 6 | 1300 | 121 | ||
GenBank mRNA | AF259898.1 | - | 6 - 7 | 786 | 121 | |
AF294257.1 | - | 6 - 7 | 1014 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
