Assay Details
Gene Symbol: Slc9a3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053811.3 | NP_446263.2 | 2 - 3 | 484 | 130 | |
GenBank mRNA | AF259898.1 | - | 2 - 3 | 343 | 130 | |
CK840521.1 | - | 2 - 3 | 271 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
