Assay Details
Gene Symbol: Hexdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142562.2 | NP_001136034.2 | 3 - 4 | 317 | 66 | |
XM_006247860.2 | XP_006247922.1 | 2 - 3 | 244 | 66 | ||
XM_008768499.1 | XP_008766721.1 | 3 - 4 | 336 | 66 | ||
GenBank mRNA | BC158670.1 | - | 2 - 3 | 254 | 66 | |
DN931298.1 | - | 3 - 4 | 347 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
