Assay Details
Gene Symbol: Hexdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142562.2 | NP_001136034.2 | 12 - 13 | 1376 | 126 | |
XM_006247860.2 | XP_006247922.1 | 11 - 12 | 1303 | 126 | ||
XM_008768499.1 | XP_008766721.1 | 12 - 13 | 1395 | 126 | ||
GenBank mRNA | BC158670.1 | - | 11 - 12 | 1313 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
