Assay Details
Gene Symbol: Pgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247855.2 | XP_006247917.1 | 7 - 8 | 1417 | 89 | |
XM_221142.8 | XP_221142.5 | 7 - 8 | 1417 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
