Assay Details
Gene Symbol: Psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134458.2 | NP_604453.2 | 2 - 3 | 158 | 68 | |
XM_006247249.2 | XP_006247311.1 | 2 - 3 | 1010 | 68 | ||
GenBank mRNA | AF352812.1 | - | 2 - 3 | 158 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
