Assay Details
Gene Symbol: Psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134458.2 | NP_604453.2 | 7 - 8 | 617 | 83 | |
GenBank mRNA | CK840952.1 | - | 1 - 2 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
