Assay Details
Gene Symbol: Psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134458.2 | NP_604453.2 | 1 - 2 | 59 | 81 | |
XM_006247249.2 | XP_006247311.1 | 1 - 2 | 911 | 81 | ||
GenBank mRNA | AF352812.1 | - | 1 - 2 | 59 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF352812.1, NM_134458.2, XM_006247249.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
