Assay Details
Gene Symbol: P3h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021581.1 | NP_067592.1 | 7 - 8 | 1225 | 80 | |
XM_006247414.2 | XP_006247476.1 | 7 - 8 | 1530 | 80 | ||
GenBank mRNA | FQ227296.1 | - | 7 - 8 | 1332 | 80 | |
X65454.1 | - | 7 - 8 | 1225 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
