Assay Details
Gene Symbol: Pipox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012009.1 | NP_001012009.1 | 2 - 3 | 266 | 104 | |
XM_006246958.1 | XP_006247020.1 | 3 - 4 | 402 | 104 | ||
XM_006246959.1 | XP_006247021.1 | 2 - 3 | 590 | 104 | ||
GenBank mRNA | BC088249.1 | - | 2 - 3 | 266 | 7370364 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
