Assay Details
Gene Symbol: Prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013181.1 | NP_037313.1 | 1 - 2 | 88 | 75 | |
GenBank mRNA | M17086.1 | - | 1 - 2 | 88 | 75 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the M17086.1, NM_013181.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
