Assay Details
Gene Symbol: Jmjd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247806.1 | XP_006247868.1 | 5 - 6 | 1107 | 97 | |
GenBank mRNA | BM391923.1 | - | 2 - 3 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
