Assay Details
Gene Symbol: Nxf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002730283.4 | XP_002730329.2 | 15 - 16 | 1384 | 119 | |
XM_006257244.2 | XP_006257306.1 | 15 - 16 | 1304 | 119 | ||
XM_006257245.2 | XP_006257307.1 | 15 - 16 | 1297 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
