Assay Details
Gene Symbol: Xrcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006999.1 | NP_001007000.1 | 7 - 8 | 899 | 108 | |
XM_006231751.1 | XP_006231813.1 | 7 - 8 | 1068 | 108 | ||
XM_008760636.1 | XP_008758858.1 | 7 - 8 | 1030 | 108 | ||
XM_008760637.1 | XP_008758859.1 | 7 - 8 | 1095 | 108 | ||
GenBank mRNA | BC083848.1 | - | 7 - 8 | 899 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
pro-B cell differentiation
DNA ligation
double-strand break repair
double-strand break repair via nonhomologous end joining
central nervous system development
response to X-ray
response to ionizing radiation
response to gamma radiation
T cell differentiation in thymus
V(D)J recombination
immunoglobulin V(D)J recombination
negative regulation of neuron apoptotic process
isotype switching
positive regulation of fibroblast proliferation
positive regulation of neurogenesis
DNA ligation involved in DNA repair
positive regulation of ligase activity
cellular response to lithium ion
molecular_function
DNA binding
protein C-terminus binding
ligase activity
in utero embryonic development
pro-B cell differentiation
DNA ligation
double-strand break repair
double-strand break repair via nonhomologous end joining
central nervous system development
response to X-ray
response to ionizing radiation
response to gamma radiation
T cell differentiation in thymus
V(D)J recombination
immunoglobulin V(D)J recombination
negative regulation of neuron apoptotic process
isotype switching
positive regulation of fibroblast proliferation
positive regulation of neurogenesis
DNA ligation involved in DNA repair
positive regulation of ligase activity
cellular response to lithium ion
molecular_function
DNA binding
protein C-terminus binding
ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P163685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95324 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P163685 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P163685 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P163685 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P163685 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4377 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91474 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35010 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69070 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76879 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26654 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34429 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40788 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31758 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64049 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44655 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76701 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20553 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41937 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103487 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48063 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10820307 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820304 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820302 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820305 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820303 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820306 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820301 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10820300 | - | - |
Rat Genome 230 2.0 Array | 1374304_at | - | - |
Rat Genome 230 2.0 Array | 1374304_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374304_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374304_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200017912.rn.1 | TC0200002573.rn.1 | RTA TC |
Rat Expression Set 230 | 1374304_at | - | - |
Rat Expression Set 230 | 1374304_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820300 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21464712 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353934 | - | - |
RatRef-12 Expression BeadChip | ILMN_48210 | - | - |
