Assay Details
Gene Symbol: Emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053719.1 | NP_446171.1 | 3 - 4 | 477 | 142 | |
GenBank mRNA | AJ009698.1 | - | 3 - 4 | 477 | 142 | |
BC061846.1 | - | 3 - 4 | 479 | 5621074 | 142 | |
FQ213962.1 | - | 3 - 4 | 490 | 142 | ||
FQ228768.1 | - | 3 - 4 | 490 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
