Assay Details
Gene Symbol: Rnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031097.1 | NP_112359.1 | 4 - 5 | 854 | 104 | |
GenBank mRNA | AY724503.1 | - | 4 - 5 | 1029 | 104 | |
BC061718.1 | - | 4 - 5 | 887 | 5598331 | 104 | |
D87515.1 | - | 4 - 5 | 851 | 104 | ||
U61696.1 | - | 4 - 5 | 854 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
