Assay Details
Gene Symbol: Rnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031097.1 | NP_112359.1 | 7 - 8 | 1318 | 140 | |
GenBank mRNA | AY724503.1 | - | 7 - 8 | 1493 | 140 | |
BC061718.1 | - | 7 - 8 | 1351 | 5598331 | 140 | |
D87515.1 | - | 7 - 8 | 1315 | 140 | ||
U61696.1 | - | 7 - 8 | 1318 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
