Assay Details
Gene Symbol: Rgs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EF568934.1 | - | 1 - 2 | 107 | 114 | |
FQ234716.1 | - | 1 - 2 | 160 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the EF568934.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
