Assay Details
Gene Symbol: Eif1ad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008305.1 | NP_001008306.1 | 4 - 5 | 838 | 92 | |
GenBank mRNA | BC085818.1 | - | 4 - 5 | 838 | 7124507 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
