Assay Details
Gene Symbol: Paox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106311.1 | NP_001099781.1 | 3 - 4 | 735 | 98 | |
XM_006230524.2 | XP_006230586.2 | 2 - 3 | 710 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
