Assay Details
Gene Symbol: Rab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109979.2 | NP_001103449.1 | 91 | 63 | ||
GenBank mRNA | BC085118.1 | - | 58 | 63 | ||
DN934711.1 | - | 109 | 63 | |||
EV762710.1 | - | 110 | 63 | |||
X13905.1 | - | 31 | 63 |
Gene Symbol: LOC102555167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760197.1 | XP_008758419.1 | 1 | 59 | 63 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the X13905.1, XM_008760197.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
