Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191917.1 | NP_001178846.1 | 37 - 38 | 5424 | 63 | |
XM_006231332.2 | XP_006231394.1 | 37 - 38 | 5710 | 63 | ||
XM_006231334.1 | XP_006231396.1 | 34 - 35 | 5086 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
