Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191917.1 | NP_001178846.1 | 4 - 5 | 417 | 70 | |
XM_006231332.2 | XP_006231394.1 | 4 - 5 | 703 | 70 | ||
XM_006231334.1 | XP_006231396.1 | 1 - 2 | 75 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006231334.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
