Assay Details
Gene Symbol: Capns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017118.1 | NP_058814.1 | 2 - 3 | 266 | 75 | |
GenBank mRNA | BC098068.1 | - | 2 - 3 | 266 | 75 | |
U53859.1 | - | 1 - 2 | 202 | 75 |
Gene Symbol: LOC100911363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748836.3 | XP_003748884.1 | 2 - 3 | 315 | 75 | |
XM_008759263.1 | XP_008757485.1 | 2 - 3 | 517 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
