Assay Details
Gene Symbol: Capns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017118.1 | NP_058814.1 | 8 - 9 | 781 | 86 | |
GenBank mRNA | BC098068.1 | - | 8 - 9 | 781 | 86 | |
FQ229527.1 | - | 464 | 86 | |||
U10861.1 | - | 470 | 86 | |||
U53859.1 | - | 7 - 8 | 717 | 86 |
Gene Symbol: LOC100912380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228796.2 | XP_006228858.2 | 557 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
