Assay Details
Gene Symbol: Nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031626.1 | NP_113814.1 | 3 - 4 | 294 | 87 | |
GenBank mRNA | BC157812.1 | - | 3 - 4 | 273 | 87 | |
U14533.1 | - | 3 - 4 | 294 | 87 | ||
U20389.1 | - | 3 - 4 | 229 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
