Assay Details
Gene Symbol: Ifnar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105893.1 | NP_001099363.2 | 10 - 11 | 1699 | 120 | |
NM_001271316.1 | NP_001258245.1 | 11 - 12 | 1889 | 120 | ||
GenBank mRNA | FQ210936.1 | - | 11 - 12 | 1689 | 120 | |
FQ218374.1 | - | 2 - 3 | 320 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P925990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76328 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P925990 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P925990 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P925990 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16946 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P925990 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10750201 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750202 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750207 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750209 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750204 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750206 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750205 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750210 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750203 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750208 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10750200 | - | - |
Rat Genome 230 2.0 Array | 1382769_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391677_at | - | - |
Rat Genome 230 2.0 Array | 1398055_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379298_at | - | - |
Rat Genome 230 2.0 Array | 1379298_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375696_at | - | - |
Rat Genome 230 2.0 Array | 1391677_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398523_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1398055_at | - | - |
Rat Genome 230 2.0 Array | 1395364_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382769_at | - | - |
Rat Genome 230 2.0 Array | 1395364_at | - | - |
Rat Genome 230 2.0 Array | 1375696_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398523_a_at | - | - |
Affymetrix HT Rat Focus Array | 1382769_at | - | - |
Affymetrix HT Rat Focus Array | 1379298_at | - | - |
Affymetrix HT Rat Focus Array | 1391677_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398523_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1379298_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398055_at | - | - |
Affymetrix HT Rat Focus Array | 1375696_at | - | - |
Affymetrix HT Rat Focus Array | 1375696_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398523_a_at | - | - |
Affymetrix HT Rat Focus Array | 1398055_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391677_at | - | - |
Affymetrix HT Rat Focus Array | 1382769_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395364_at | - | - |
Affymetrix HT Rat Focus Array | 1395364_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100000976.rn.1 | TC1100000250.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100000975.rn.1 | TC1100000250.rn.1 | RTA TC |
Rat Expression Set 230 | 1379298_at | - | - |
Rat Expression Set 230 | 1398523_PM_a_at | - | - |
Rat Expression Set 230 | 1391677_PM_at | - | - |
Rat Expression Set 230 | 1375696_PM_at | - | - |
Rat Expression Set 230 | 1391677_at | - | - |
Rat Expression Set 230 | 1375696_at | - | - |
Rat Expression Set 230 | 1379298_PM_at | - | - |
Rat Expression Set 230 | 1398523_a_at | - | - |
Rat Expression Set 230 | 1398055_at | - | - |
Rat Expression Set 230 | 1395364_PM_at | - | - |
Rat Expression Set 230 | 1395364_at | - | - |
Rat Expression Set 230 | 1398055_PM_at | - | - |
Rat Expression Set 230 | 1382769_at | - | - |
Rat Expression Set 230 | 1382769_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750200 | - | - |
Rat Genome U34 Set | rc_AA799711_at | - | - |
Rat Genome U34 Set | rc_AI045696_at | - | - |
Rat Genome U34 Set | rc_AA996880_at | - | - |
Rat Genome U34 Set | rc_AA899403_g_at | - | - |
Rat Genome U34 Set | rc_AA900618_at | - | - |
Rat Genome U34 Set | rc_AI007872_at | - | - |
Rat Genome U34 Set | rc_AA899403_at | - | - |
Rat Genome U34 Set | rc_AI178178_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21990077 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367808 | - | - |
