Assay Details
Gene Symbol: Ifnar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105893.1 | NP_001099363.2 | 1 - 2 | 355 | 109 | |
NM_001271316.1 | NP_001258245.1 | 1 - 2 | 355 | 109 | ||
GenBank mRNA | FQ210936.1 | - | 1 - 2 | 155 | 109 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001271316.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
