Assay Details
Gene Symbol: Ifnar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105893.1 | NP_001099363.2 | 7 - 8 | 1256 | 164 | |
NM_001271316.1 | NP_001258245.1 | 8 - 9 | 1446 | 164 | ||
GenBank mRNA | FQ210936.1 | - | 8 - 9 | 1246 | 164 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
