Assay Details
Gene Symbol: Ifnar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105893.1 | NP_001099363.2 | 8 - 9 | 1399 | 119 | |
NM_001271316.1 | NP_001258245.1 | 9 - 10 | 1589 | 119 | ||
GenBank mRNA | FQ210936.1 | - | 9 - 10 | 1389 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
