Assay Details
Gene Symbol: Clic6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176078.2 | NP_788267.1 | 5 - 6 | 1987 | 126 | |
GenBank mRNA | AY191249.1 | - | 5 - 6 | 1987 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of ion transmembrane transport
chloride transmembrane transport
voltage-gated ion channel activity
chloride channel activity
protein C-terminus binding
D2 dopamine receptor binding
D3 dopamine receptor binding
D4 dopamine receptor binding
protein homodimerization activity
dopamine receptor binding
regulation of ion transmembrane transport
chloride transmembrane transport
voltage-gated ion channel activity
chloride channel activity
protein C-terminus binding
D2 dopamine receptor binding
D3 dopamine receptor binding
D4 dopamine receptor binding
protein homodimerization activity
dopamine receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P449623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27208 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P449623 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P449623 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P449623 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P449623 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92887 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22099 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21823 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10750299 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750297 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750304 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750300 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750298 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750302 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750301 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750305 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750303 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10750296 | - | - |
Affymetrix Rat Clariom S Assay | PSR1100001245.rn.1 | TC1100000285.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10750296 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21215032 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371422 | - | - |
RatRef-12 Expression BeadChip | ILMN_50949 | - | - |
