Assay Details
Gene Symbol: Ifnar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248107.2 | XP_006248169.1 | 5 - 6 | 688 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
