Assay Details
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105897.2 | NP_001099367.2 | 2 - 3 | 284 | 66 | |
GenBank mRNA | DY312304.1 | - | 2 - 3 | 284 | 66 | |
FQ225223.1 | - | 2 - 3 | 257 | 66 | ||
FQ226011.1 | - | 2 - 3 | 257 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
