Assay Details
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105897.2 | NP_001099367.2 | 5 - 6 | 691 | 114 | |
GenBank mRNA | CV076369.1 | - | 2 - 3 | 269 | 114 | |
FQ225223.1 | - | 5 - 6 | 664 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
