Assay Details
Gene Symbol: Nrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100560.1 | NP_001094030.1 | 2 - 3 | 254 | 128 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001100560.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
