Assay Details
Gene Symbol: Map3k7cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013979.1 | NP_001014001.1 | 1 - 2 | 238 | 107 | |
GenBank mRNA | BC076389.1 | - | 1 - 2 | 238 | 107 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC076389.1, NM_001013979.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
