Assay Details
Gene Symbol: Map3k7cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013979.1 | NP_001014001.1 | 2 - 3 | 341 | 86 | |
XM_006248016.2 | XP_006248078.1 | 3 - 4 | 1053 | 86 | ||
GenBank mRNA | BC076389.1 | - | 2 - 3 | 341 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
