Assay Details
Gene Symbol: Itgb1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239979.2 | XP_006240041.1 | 1 - 2 | 131 | 95 | |
GenBank mRNA | FQ218542.1 | - | 1 - 2 | 92 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006239979.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
