Assay Details
Gene Symbol: Itgb1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106719.2 | NP_001100189.1 | 3 - 4 | 341 | 80 | |
XM_006239979.2 | XP_006240041.1 | 2 - 3 | 257 | 80 | ||
XM_006239980.2 | XP_006240042.1 | 3 - 4 | 286 | 80 | ||
XM_006239981.2 | XP_006240043.1 | 2 - 3 | 468 | 80 | ||
GenBank mRNA | BC169104.1 | - | 3 - 4 | 341 | 80 | |
FQ215419.1 | - | 2 - 3 | 176 | 80 | ||
FQ218542.1 | - | 2 - 3 | 218 | 80 | ||
FQ229091.1 | - | 3 - 4 | 344 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
